missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Piperazinecarboxylic Acid Dihydrochloride 98.0+%, TCI America™
$96.94 - $279.60
Chemical Identifiers
| CAS | 3022-15-9 |
|---|---|
| Molecular Formula | C5H12Cl2N2O2 |
| Molecular Weight (g/mol) | 203.063 |
| MDL Number | MFCD00044803 |
| InChI Key | WNSDZBQLMGKPQS-UHFFFAOYSA-N |
| Synonym | piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride |
| PubChem CID | 2723757 |
| IUPAC Name | piperazine-2-carboxylic acid;dihydrochloride |
| SMILES | C1CNC(CN1)C(=O)O.Cl.Cl |
Chemical Identifiers
| 3022-15-9 | |
| 203.063 | |
| WNSDZBQLMGKPQS-UHFFFAOYSA-N | |
| 2723757 | |
| C1CNC(CN1)C(=O)O.Cl.Cl |
| C5H12Cl2N2O2 | |
| MFCD00044803 | |
| piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride | |
| piperazine-2-carboxylic acid;dihydrochloride |
Specifications
| 3022-15-9 | |
| C5H12Cl2N2O2 | |
| 5 g | |
| WNSDZBQLMGKPQS-UHFFFAOYSA-N | |
| piperazine-2-carboxylic acid;dihydrochloride | |
| 2723757 | |
| ≥98.0% (T) | |
| 2-Piperazinecarboxylic Acid Dihydrochloride |
| 258°C | |
| MFCD00044803 | |
| piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride | |
| C1CNC(CN1)C(=O)O.Cl.Cl | |
| 203.063 | |
| 203.06 | |
| Crystalline Powder |
Safety and Handling
TSCA : No