accessibility menu, dialog, popup

UserName

CAS RN 3022-15-9

OO(R)NH2H2N

CAS RN 3022-15-9

IUPAC Name: (3R)-piperazine-1,4-diium-3-carboxylate
Synonyms: (3R)-piperazine-1,4-diium-3-carboxylate
Molecular Weight (g/mol): 131.15
Molecular Formula: C5H11N2O2
InChi Key: JSSXHAMIXJGYCS-SCSAIBSYSA-O
SMILES: [O-]C(=O)[C@H]1C[NH2+]CC[NH2+]1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)