missing translation for 'onlineSavingsMsg'
Learn More

2-Picolinic acid, 99%

Catalog No. p-7022126
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1474922 100 g
AAA1474936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1474922 Supplier Thermo Scientific Chemicals Supplier No. A1474922
Only null left

CAS: 98-98-6 | C6H5NO2 | 123.111 g/mol

2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.

Solubility
Soluble in water. Also soluble in glacial acetic acid, ethanol and methanol. Insoluble in chloroform.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents and bases.

Chemical Identifiers

CAS 98-98-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00006293
InChI Key SIOXPEMLGUPBBT-UHFFFAOYSA-N
Synonym picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique
PubChem CID 1018
ChEBI CHEBI:28747
IUPAC Name pyridine-2-carboxylic acid
SMILES C1=CC=NC(=C1)C(=O)O

Specifications

Melting Point 135°C to 139°C
Flash Point 139°C (232°F)
Odor Odorless
Quantity 100 g
Beilstein 109595
Merck Index 14,7403
Solubility Information Soluble in water. Also soluble in glacial acetic acid,ethanol and methanol. Insoluble in chloroform.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material 2-Picolinic acid
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 202-719-7
RTECSNumber TJ7344000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only