Learn More
2-Picoline N-oxide, 98%
CAS: 931-19-1 | C6H7NO | 109.128 g/mol
Supplier: Thermo Scientific Chemicals L0448822
Description
2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under dry inert gas. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates.
Specifications
| 2-Picoline N-oxide | |
| 46°C to 50°C | |
| 143°C (289°F) | |
| MFCD00006199 | |
| 106916 | |
| 2-picoline-n-oxide, 2-methylpyridine 1-oxide, 2-methylpyridine n-oxide, picoline n-oxide, 2-picoline n-oxide, methylpyridine 1-oxide, alpha-picoline n-oxide, pyridine, 2-methyl-, 1-oxide, 2-picoline, 1-oxide, picoline 1-oxide | |
| CFZKDDTWZYUZKS-UHFFFAOYSA-N | |
| 2-methyl-1-oxidopyridin-1-ium | |
| 13602 | |
| 109.13 |
| 931-19-1 | |
| 122°C (8 mmHg) | |
| C6H7NO | |
| 100 g | |
| Hygroscopic | |
| Soluble in water. | |
| CC1=CC=CC=[N+]1[O-] | |
| 109.128 | |
| CHEBI:35578 | |
| 98% |
Chemical Identifiers
| 931-19-1 | |
| 109.128 | |
| CFZKDDTWZYUZKS-UHFFFAOYSA-N | |
| 13602 | |
| 2-methyl-1-oxidopyridin-1-ium |
| C6H7NO | |
| MFCD00006199 | |
| 2-picoline-n-oxide, 2-methylpyridine 1-oxide, 2-methylpyridine n-oxide, picoline n-oxide, 2-picoline n-oxide, methylpyridine 1-oxide, alpha-picoline n-oxide, pyridine, 2-methyl-, 1-oxide, 2-picoline, 1-oxide, picoline 1-oxide | |
| CHEBI:35578 | |
| CC1=CC=CC=[N+]1[O-] |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-230-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only