missing translation for 'onlineSavingsMsg'
Learn More

2-Picoline, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111016561
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016561 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016561 missing translation for 'mfr' Merck Emd Millipore missing translation for 'supplierNo' 74350500MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Ignition Point 995°F
CAS 109-06-8
Vapor Pressure 10 mmHg (24.4°C)
Vapor Density 3.2 (vs. air)
Density 0.943 g/mL (at 25°C)
Flash Point 29°C - Closed Cup
Physical Form Neat
Linear Formula C6H7 N
MDL Number MFCD00006332
Melting Point -70°C
Boiling Point 128°C to 129°C
Synonym alpha-Picoline; 2-Methylpyridine; NSC 3409
Type Analytical Standard
Quantity 500 mg
Formula Weight 93.13
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less