missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthiomethylbenzoic Acid, TCI America™

Catalog No. P13605G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13605G Supplier TCI America Supplier No. P13605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1699-03-2
Molecular Formula C14H11O2S
Molecular Weight (g/mol) 243.30
MDL Number MFCD00089444
InChI Key QXPRAGVOCILNHG-UHFFFAOYSA-M
Synonym 2-phenylthio methyl benzoic acid, 2-phenylthiomethylbenzoic acid, 2-phenylsulfanyl methyl benzoic acid, 2-phenylthiomethyl benzoic acid, benzoic acid, 2-phenylthio methyl, acmc-1bs3y, 2-phenylthiomethylbenzoicacid, o-phenylthiomethylbenzoic acid, 2-phenylmercaptomethylbenzoic acid
PubChem CID 74337
IUPAC Name 2-[(phenylsulfanyl)methyl]benzoate
SMILES [O-]C(=O)C1=CC=CC=C1CSC1=CC=CC=C1

Specifications

Melting Point 111°C
Color Yellow
Quantity 5 g
Formula Weight 244.31
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylthiomethylbenzoic Acid