missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthioacetamide 98.0+%, TCI America™

Catalog No. P21271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21271G 1 g
P21275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21271G Supplier TCI America Supplier No. P21271G

Chemical Identifiers

CAS 645-54-5
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.227
MDL Number MFCD00022177
InChI Key CJXBHFANXQMZBF-UHFFFAOYSA-N
Synonym 2-phenylthioacetamide, benzeneethanethioamide, phenylthioacetamide, acetamide, 2-phenylthio, benzeneethanimidothioic acid, phenylacetothioamide, 1-amino-2-phenylethane-1-thione, a-phenylthioacetamide, 2-phenyl-thioacetamide, alpha-phenylthioacetamide
PubChem CID 731368
IUPAC Name 2-phenylethanethioamide
SMILES C1=CC=C(C=C1)CC(=S)N

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Formula Weight 151.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylthioacetamide