missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenylthio)thiophene, 97+%

CAS: 16718-12-0 | C10H8S2 | 192.29 g/mol

Supplier:  Thermo Scientific Chemicals A1370806

 View more versions of this product

Catalog No. AAA1370806

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(Phenylthio)thiophene
109°C to 110°C (0.4 mmHg)
MFCD00052141
2-phenylthio thiophene, 2-phenylsulfanyl thiophene, 2-phenylthiothiophene, phenylthiophenyl sulfide, acmc-1bvxh, 2-thienyl phenylsulfide, thiophene,2-phenylthio, thiophene, 2-phenylthio
S(C1=CC=CS1)C1=CC=CC=C1
192.29
192.3
16718-12-0
C10H8S2
5 g
JQTBWKNYWACCRU-UHFFFAOYSA-N
2-phenylsulfanylthiophene
2778915
≥97%

Chemical Identifiers

16718-12-0
192.29
JQTBWKNYWACCRU-UHFFFAOYSA-N
2-phenylsulfanylthiophene
C10H8S2
MFCD00052141
2778915
S(C1=CC=CS1)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only