missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenylthio)thiophene, 97+%

Catalog No. AAA1370803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1370803 1 g
AAA1370806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1370803 Supplier Thermo Scientific Chemicals Supplier No. A1370803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16718-12-0 | C10H8S2 | 192.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16718-12-0
Molecular Formula C10H8S2
Molecular Weight (g/mol) 192.29
MDL Number MFCD00052141
InChI Key JQTBWKNYWACCRU-UHFFFAOYSA-N
PubChem CID 2778915
IUPAC Name 2-phenylsulfanylthiophene
SMILES S(C1=CC=CS1)C1=CC=CC=C1

Specifications

Boiling Point 109°C to 110°C (0.4 mmHg)
Quantity 1 g
Synonym 2-phenylthio thiophene, 2-phenylsulfanyl thiophene, 2-phenylthiothiophene, phenylthiophenyl sulfide, acmc-1bvxh, 2-thienyl phenylsulfide, thiophene,2-phenylthio, thiophene, 2-phenylthio
Formula Weight 192.3
Percent Purity ≥97%
Chemical Name or Material 2-(Phenylthio)thiophene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only