missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylsulfonylacetophenone 98.0+%, TCI America™

Catalog No. P08285G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P08285G Supplier TCI America Supplier No. P08285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3406-03-9
Molecular Formula C14H12O3S
Molecular Weight (g/mol) 260.31
MDL Number MFCD00025043
InChI Key DREVPGKOIZVPQV-UHFFFAOYSA-N
Synonym 2-phenylsulfonyl acetophenone, 2-phenylsulfonylacetophenone, 1-phenyl-2-phenylsulfonyl ethanone, acetophenone, 2-phenylsulfonyl, 2-benzenesulfonyl-1-phenylethanone, ethanone, 1-phenyl-2-phenylsulfonyl, 1-phenyl-2-phenylsulphonyl ethan-1-one, alpha-phenylsulfonylacetophenone, 2-benzenesulfonyl-1-phenylethan-1-one, 1-phenyl-2-phenylsulfonyl ethan-1-one
PubChem CID 76949
IUPAC Name 2-(benzenesulfonyl)-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
Formula Weight 260.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylsulfonylacetophenone