missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpropylene Oxide 95.0+%, TCI America™

Catalog No. p08335ml
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
P08335ML 5 mL
1 options

Catalog No. P08335ML

Supplier: TCI America P08335ML

Only null left

Chemical Identifiers

CAS 2085-88-3
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00059156
InChI Key MRXPNWXSFCODDY-UHFFFAOYNA-N
Synonym 1,2-Epoxy-2-phenylpropane, alpha-Methylstyrene Oxide
PubChem CID 16398
IUPAC Name 2-methyl-2-phenyloxirane
SMILES CC1(CO1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 mL
Formula Weight 134.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylpropylene Oxide
RTECSNumber RR0532400
TSCA No