accessibility menu, dialog, popup

UserName

H3CO

CAS RN 2085-88-3

IUPAC Name: 2-methyl-2-phenyloxirane
Synonyms: 2-methyl-2-phenyloxirane
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: MRXPNWXSFCODDY-UHFFFAOYNA-N
SMILES: CC1(CO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results