missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylisobutyric Acid 98.0+%, TCI America™

Catalog No. p-7135124
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P140925G 25 g
P1409250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P140925G Supplier TCI America Supplier No. P140925G
Only null left

Chemical Identifiers

CAS 826-55-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00014332
InChI Key YYEROYLAYAVZNW-UHFFFAOYSA-N
Synonym 2-methyl-2-phenylpropionic acid, 2-phenylisobutyric acid, alpha,alpha-dimethylphenylacetic acid, unii-dr8huh2gyk, dimethylphenylacetic acid, dr8huh2gyk, 2-methyl-2-phenyl-propionic acid, hydratropic acid, .alpha.-methyl, benzeneacetic acid,.alpha.-dimethyl, propionic acid, 2-methyl-2-phenyl
PubChem CID 13222
IUPAC Name 2-methyl-2-phenylpropanoic acid
SMILES CC(C)(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylisobutyric Acid