missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylindole 98.0+%, TCI America™

Catalog No. P018825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P018825G 25 g
P0188250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P018825G Supplier TCI America Supplier No. P018825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 948-65-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00005608
InChI Key KLLLJCACIRKBDT-UHFFFAOYSA-N
Synonym 2-phenylindole, 1h-indole, 2-phenyl, indole, 2-phenyl, stabilizer i, alpha-phenylindole, 2-phenyl indole, phenylindole, unii-mqd44hv3p1, .alpha.-phenylindole, mqd44hv3p1
PubChem CID 13698
IUPAC Name 2-phenyl-1H-indole
SMILES C1=CC=C(C=C1)C2=CC3=CC=CC=C3N2

Specifications

Melting Point 192°C
Color Yellow
Quantity 25 g
Formula Weight 193.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylindole
EINECSNumber (5)-0137
TSCA Yes