missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylindole, 95%

Catalog No. AAB2367430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2367430 250 g
AAB2367418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2367430 Supplier Thermo Scientific Chemicals Supplier No. B2367430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 948-65-2 | C14H11N | 193.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 948-65-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00005608
InChI Key KLLLJCACIRKBDT-UHFFFAOYSA-N
Synonym 2-phenylindole, 1h-indole, 2-phenyl, indole, 2-phenyl, stabilizer i, alpha-phenylindole, 2-phenyl indole, phenylindole, unii-mqd44hv3p1, .alpha.-phenylindole, mqd44hv3p1
PubChem CID 13698
IUPAC Name 2-phenyl-1H-indole
SMILES C1=CC=C(C=C1)C2=CC3=CC=CC=C3N2

Specifications

Melting Point 189°C to 190°C
Boiling Point 250°C (10 mmHg)
Quantity 250 g
Formula Weight 193.25
Percent Purity 95%
Chemical Name or Material 2-Phenylindole
Hazard Category Danger
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 213-436-3
RTECSNumber NM1272500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only