missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylglycinol 98.0+%, TCI America™

Supplier:  TCI America P179225G

 View more versions of this product

Catalog No. P179225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Phenylglycinol
51°C
106°C
MFCD00130145
2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
NC(CO)C1=CC=CC=C1
137.18
137.18
Crystalline Powder
7568-92-5
White-Yellow
C8H11NO
25 g
IJXJGQCXFSSHNL-UHFFFAOYNA-N
2-amino-2-phenylethan-1-ol
92466
≥98.0% (GC,T)

Chemical Identifiers

7568-92-5
137.18
IJXJGQCXFSSHNL-UHFFFAOYNA-N
92466
NC(CO)C1=CC=CC=C1
C8H11NO
MFCD00130145
2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
2-amino-2-phenylethan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : SG7600000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions