missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylglycinol 98.0+%, TCI America™
$143.54 - $566.67
Chemical Identifiers
| CAS | 7568-92-5 |
|---|---|
| Molecular Formula | C8H11NO |
| Molecular Weight (g/mol) | 137.18 |
| MDL Number | MFCD00130145 |
| InChI Key | IJXJGQCXFSSHNL-UHFFFAOYNA-N |
| Synonym | 2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino |
| PubChem CID | 92466 |
| IUPAC Name | 2-amino-2-phenylethan-1-ol |
| SMILES | NC(CO)C1=CC=CC=C1 |
Chemical Identifiers
| 7568-92-5 | |
| 137.18 | |
| IJXJGQCXFSSHNL-UHFFFAOYNA-N | |
| 92466 | |
| NC(CO)C1=CC=CC=C1 |
| C8H11NO | |
| MFCD00130145 | |
| 2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino | |
| 2-amino-2-phenylethan-1-ol |
Specifications
| 7568-92-5 | |
| White-Yellow | |
| C8H11NO | |
| 5 g | |
| IJXJGQCXFSSHNL-UHFFFAOYNA-N | |
| 2-amino-2-phenylethan-1-ol | |
| 92466 | |
| ≥98.0% (GC,T) | |
| 2-Phenylglycinol |
| 51°C | |
| 106°C | |
| MFCD00130145 | |
| 2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino | |
| NC(CO)C1=CC=CC=C1 | |
| 137.18 | |
| 137.18 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : SG7600000
TSCA : No
Recommended Storage : Refrigerator