missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethylamine Hydrobromide 98.0+%, TCI America™

Catalog No. P23881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23881G 1 g
P23885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23881G Supplier TCI America Supplier No. P23881G

Chemical Identifiers

CAS 53916-94-2
Molecular Formula C8H12BrN
Molecular Weight (g/mol) 202.095
InChI Key IRAGENYJMTVCCV-UHFFFAOYSA-N
Synonym Phenethylamine Hydrobromide, 2-Phenylethylammonium Bromide
PubChem CID 70441016
IUPAC Name 2-phenylethanamine;hydrobromide
SMILES C1=CC=C(C=C1)CCN.Br

Specifications

Melting Point 259°C
Color White
Quantity 1 g
Formula Weight 202.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylethylamine Hydrobromide