Learn More
2-Phenylethylamine, 99%
CAS: 64-04-0 | C8H11N | 121.183 g/mol
$45.81 - $361.51
Chemical Identifiers
CAS | 64-04-0 |
---|---|
Molecular Formula | C8H11N |
Molecular Weight (g/mol) | 121.183 |
MDL Number | MFCD00008184 |
InChI Key | BHHGXPLMPWCGHP-UHFFFAOYSA-N |
Synonym | phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzeneShow More |
PubChem CID | 1001 |
ChEBI | CHEBI:18397 |
IUPAC Name | 2-phenylethanamine |
SMILES | C1=CC=C(C=C1)CCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA13402AD
|
Thermo Scientific Chemicals
A13402AD |
50 mL |
Each for $45.81
|
|
||||||
AAA13402AK
|
Thermo Scientific Chemicals
A13402AK |
250 mL |
Each for $112.03
|
|
||||||
AAA13402AU
|
Thermo Scientific Chemicals
A13402AU |
1000 mL |
Each for $361.51
|
|
||||||
Description
Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Store under inert gas. Keep away from acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide (CO2).
Chemical Identifiers
64-04-0 | |
121.183 | |
BHHGXPLMPWCGHP-UHFFFAOYSA-N | |
1001 | |
2-phenylethanamine |
C8H11N | |
MFCD00008184 | |
phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzeneShow More | |
CHEBI:18397 | |
C1=CC=C(C=C1)CCN |
Specifications
64-04-0 | |
0.964 | |
200°C to 202°C | |
Amine-like | |
1.5335 | |
MFCD00008184 | |
UN2922 | |
Air Sensitive | |
phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene | |
BHHGXPLMPWCGHP-UHFFFAOYSA-N | |
2-phenylethanamine | |
1001 | |
121.18 | |
2-Phenylethylamine |
-60°C | |
11.5 | |
81°C (178°F) | |
C8H11N | |
C6H5(CH2)2NH2 | |
50 mL | |
507488 | |
14,7225 | |
It is soluble in water. | |
C1=CC=C(C=C1)CCN | |
121.183 | |
CHEBI:18397 | |
99% |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H301-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 200-574-4
RTECSNumber : SG8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only