Learn More
2-Phenylethylamine, 99%
CAS: 64-04-0 | C8H11N | 121.183 g/mol
Supplier: Thermo Scientific Chemicals A13402AK
Description
Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Store under inert gas. Keep away from acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide (CO2).
Specifications
| 2-Phenylethylamine | |
| -60°C | |
| 11.5 | |
| 81°C (178°F) | |
| C8H11N | |
| C6H5(CH2)2NH2 | |
| 250 mL | |
| 507488 | |
| 14,7225 | |
| It is soluble in water. | |
| C1=CC=C(C=C1)CCN | |
| 121.183 | |
| CHEBI:18397 | |
| 99% |
| 64-04-0 | |
| 0.964 | |
| 200°C to 202°C | |
| Amine-like | |
| 1.5335 | |
| MFCD00008184 | |
| UN2922 | |
| Air Sensitive | |
| phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene | |
| BHHGXPLMPWCGHP-UHFFFAOYSA-N | |
| 2-phenylethanamine | |
| 1001 | |
| 121.18 |
Chemical Identifiers
| 64-04-0 | |
| 121.183 | |
| BHHGXPLMPWCGHP-UHFFFAOYSA-N | |
| 1001 | |
| 2-phenylethanamine |
| C8H11N | |
| MFCD00008184 | |
| phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene | |
| CHEBI:18397 | |
| C1=CC=C(C=C1)CCN |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H301-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 200-574-4
RTECSNumber : SG8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only