missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl isothiocyanate, 98%

Catalog No. AAA1512314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1512314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1512314 Supplier Thermo Scientific Chemicals Supplier No. A1512314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2257-09-2 | C9H9NS | 163.24 g/mol

Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

Solubility
Soluble in chloroform, ethyl acetate, and dichoromethane. Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under dry inert gas.

Chemical Identifiers

CAS 2257-09-2
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00004821
InChI Key IZJDOKYDEWTZSO-UHFFFAOYSA-N
Synonym 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
PubChem CID 16741
ChEBI CHEBI:351346
IUPAC Name 2-isothiocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=S

Specifications

Density 1.09
Boiling Point 139°C to 140°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5888
Quantity 25 g
UN Number UN2922
Beilstein 2084162
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,ethyl acetate,and dichoromethane. Reacts with water.
Formula Weight 163.24
Percent Purity 98%
Chemical Name or Material 2-Phenylethyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 218-855-5
RTECSNumber NX9115000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only