missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl Isobutyrate 98.0+%, TCI America™

Catalog No. p-7135358
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P200825ML 25 mL
P2008500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P200825ML Supplier TCI America Supplier No. P200825ML
Only null left

Chemical Identifiers

CAS 103-48-0
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.26
MDL Number MFCD00026441
InChI Key JDQVBGQWADMTAM-UHFFFAOYSA-N
Synonym Isobutyric Acid 2-Phenylethyl Ester, Phenethyl Isobutyrate, Isobutyric Acid Phenethyl Ester
PubChem CID 7655
ChEBI CHEBI:87409
IUPAC Name 2-phenylethyl 2-methylpropanoate
SMILES CC(C)C(=O)OCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 250°C
Quantity 25 mL
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylethyl Isobutyrate
EINECSNumber (3)-1040&(3)-1045
RTECSNumber NQ5435000
TSCA Yes