CAS RN 103-48-0
IUPAC Name:
2-phenylethyl 2-methylpropanoate
Synonyms:
phenethyl isobutyrate
Molecular Weight (g/mol):
192.26
Molecular Formula:
C12H16O2
InChi Key:
JDQVBGQWADMTAM-UHFFFAOYSA-N
SMILES:
CC(C)C(=O)OCCC1=CC=CC=C1
1
–
1
of
1
results
1
2-Phenylethyl Isobutyrate 98.0+%, TCI America™
CAS: 103-48-0 Molecular Formula: C12H16O2 Molecular Weight (g/mol): 192.26 MDL Number: MFCD00026441 InChI Key: JDQVBGQWADMTAM-UHFFFAOYSA-N Synonym: Isobutyric Acid 2-Phenylethyl Ester, Phenethyl Isobutyrate, Isobutyric Acid Phenethyl Ester PubChem CID: 7655 ChEBI: CHEBI:87409 IUPAC Name: 2-phenylethyl 2-methylpropanoate SMILES: CC(C)C(=O)OCCC1=CC=CC=C1