missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl Formate 95.0+%, TCI America™

Catalog No. F036325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F036325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F036325G Supplier TCI America Supplier No. F036325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104-62-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00021046
InChI Key IKDIJXDZEYHZSD-UHFFFAOYSA-N
Synonym Formic Acid Phenethyl Ester, Phenethyl Formate, Formic Acid 2-Phenylethyl Ester
PubChem CID 7711
ChEBI CHEBI:87417
IUPAC Name 2-phenylethyl formate
SMILES O=COCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 117°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylethyl Formate
EINECSNumber (3)-1040
RTECSNumber LQ9400000
TSCA Yes