accessibility menu, dialog, popup

UserName

CAS RN 104-62-1

IUPAC Name: 2-phenylethyl formate
Synonyms: phenethyl formate
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: IKDIJXDZEYHZSD-UHFFFAOYSA-N
SMILES: O=COCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results