missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl Alcohol 98.0+%, TCI America™

Catalog No. P008425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P008425ML 25 mL
P0084500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P008425ML Supplier TCI America Supplier No. P008425ML

Chemical Identifiers

CAS 60-12-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002886
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
Synonym phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
PubChem CID 6054
ChEBI CHEBI:49000
IUPAC Name 2-phenylethan-1-ol
SMILES OCCC1=CC=CC=C1

Specifications

Melting Point -27°C
Color Colorless
Boiling Point 219°C
Quantity 25 mL
UN Number 2810
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylethyl Alcohol
EINECSNumber (3)-1032
RTECSNumber SG7175000
TSCA Yes