missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanol, 98+%

CAS: 60-12-8 | C8H10O | 122.17 g/mol

Supplier:  Thermo Scientific Chemicals A152410I

 View more versions of this product

Catalog No. AAA152410I


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.

Solubility
Miscible with chloroform.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Phenylethanol
-27°C
219°C
≥98%
1.533
MFCD00002886
UN2810
14,7224
Miscible with chloroform.
OCCC1=CC=CC=C1
122.17
CHEBI:49000
≥98%
60-12-8
1.021
101°C (214°F)
C8H10O
C6H5(CH2)2OH
5000 g
1905732
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
WRMNZCZEMHIOCP-UHFFFAOYSA-N
2-phenylethan-1-ol
6054
122.17
Liquid

Chemical Identifiers

60-12-8
122.17
WRMNZCZEMHIOCP-UHFFFAOYSA-N
6054
OCCC1=CC=CC=C1
C8H10O
MFCD00002886
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
CHEBI:49000
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 200-456-2

RTECSNumber : SG7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only