Learn More
(+/-)-2-Phenylbutyric acid, 98%, Thermo Scientific Chemicals
$118.63 - $118.63
Chemical Identifiers
| CAS | 90-27-7 |
|---|---|
| Molecular Formula | C10H12O2 |
| Molecular Weight (g/mol) | 164.204 |
| MDL Number | MFCD00002667 |
| InChI Key | OFJWFSNDPCAWDK-UHFFFAOYSA-N |
| Synonym | 2-phenylbutyric acid, alpha-ethylphenylacetic acid, butyric acid, 2-phenyl, alpha-phenylbutyric acid, benzeneacetic acid, .alpha.-ethyl, alpha-phenyl butyric acid, 2-phenylbutyricacid, benzeneacetic acid, alpha-ethyl, 2-phenylburyric acid, alpha-toluic acid, alpha-ethyl |
| PubChem CID | 7012 |
| ChEBI | CHEBI:86545 |
| IUPAC Name | 2-phenylbutanoic acid |
| SMILES | CCC(C1=CC=CC=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2322222
|
Thermo Scientific Chemicals
B2322222 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 90-27-7 | |
| 164.204 | |
| OFJWFSNDPCAWDK-UHFFFAOYSA-N | |
| 7012 | |
| 2-phenylbutanoic acid |
| C10H12O2 | |
| MFCD00002667 | |
| 2-phenylbutyric acid, alpha-ethylphenylacetic acid, butyric acid, 2-phenyl, alpha-phenylbutyric acid, benzeneacetic acid, .alpha.-ethyl, alpha-phenyl butyric acid, 2-phenylbutyricacid, benzeneacetic acid, alpha-ethyl, 2-phenylburyric acid, alpha-toluic acid, alpha-ethyl | |
| CHEBI:86545 | |
| CCC(C1=CC=CC=C1)C(=O)O |
Specifications
| 90-27-7 | |
| 270°C to 272°C | |
| C10H12O2 | |
| MFCD00002667 | |
| 509876 | |
| OFJWFSNDPCAWDK-UHFFFAOYSA-N | |
| 2-phenylbutanoic acid | |
| 7012 | |
| 164.2 | |
| (+/-)-2-Phenylbutyric acid |
| 40°C to 44°C | |
| >110°C (230°F) | |
| 1.515 | |
| 100 g | |
| 2-phenylbutyric acid, alpha-ethylphenylacetic acid, butyric acid, 2-phenyl, alpha-phenylbutyric acid, benzeneacetic acid, .alpha.-ethyl, alpha-phenyl butyric acid, 2-phenylbutyricacid, benzeneacetic acid, alpha-ethyl, 2-phenylburyric acid, alpha-toluic acid, alpha-ethyl | |
| CCC(C1=CC=CC=C1)C(=O)O | |
| 164.204 | |
| CHEBI:86545 | |
| 98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 201-982-5
RTECSNumber : ET5957500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only