missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Phenylbutyric acid, 98%, Thermo Scientific Chemicals

Catalog No. AAB2322222
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2322222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2322222 Supplier Thermo Scientific Chemicals Supplier No. B2322222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90-27-7 | Molecular Formula: C10H12O2 | Molecular Weight: 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-27-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002667
InChI Key OFJWFSNDPCAWDK-UHFFFAOYSA-N
Synonym 2-phenylbutyric acid, alpha-ethylphenylacetic acid, butyric acid, 2-phenyl, alpha-phenylbutyric acid, benzeneacetic acid, .alpha.-ethyl, alpha-phenyl butyric acid, 2-phenylbutyricacid, benzeneacetic acid, alpha-ethyl, 2-phenylburyric acid, alpha-toluic acid, alpha-ethyl
PubChem CID 7012
ChEBI CHEBI:86545
IUPAC Name 2-phenylbutanoic acid
SMILES CCC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 40°C to 44°C
Boiling Point 270°C to 272°C
Flash Point >110°C (230°F)
Refractive Index 1.515
Quantity 100 g
Beilstein 509876
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material (+/-)-2-Phenylbutyric acid
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-982-5
RTECSNumber ET5957500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only