missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzoxazole, 99%

Catalog No. p-7027856
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2490603 1 g
AAB2490606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2490603 Supplier Thermo Scientific Chemicals Supplier No. B2490603
Only null left

CAS: 833-50-1 | C13H9NO | 195.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 833-50-1
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00012183
InChI Key FIISKTXZUZBTRC-UHFFFAOYSA-N
Synonym 2-phenylbenzoxazole, 2-phenylbenzo d oxazole, benzoxazole, 2-phenyl, ccris 7874, phenyl-2 benzoxazole french, 2-phenyl-benzoxazole, phenyl-2 benzoxazole, 2-phenybenzoxazole, 2-phenyl benzoxazole, 2-phenyl-benzooxazole
PubChem CID 70030
IUPAC Name 2-phenyl-1,3-benzoxazole
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3O2

Specifications

Melting Point 102°C to 104°C
Quantity 1 g
Formula Weight 195.22
Percent Purity 99%
Chemical Name or Material 2-Phenylbenzoxazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DM4865700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only