accessibility menu, dialog, popup

UserName

CAS RN 833-50-1

IUPAC Name: 2-phenyl-1,3-benzoxazole
Synonyms: benzoxazole, 2-phenyl-
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: FIISKTXZUZBTRC-UHFFFAOYSA-N
SMILES: O1C2=CC=CC=C2N=C1C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)