missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
Supplier: TCI America P185525G
Specifications
| 2-Phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| White-Yellow | |
| C12H17BO2 | |
| 25 g | |
| KKLCYBZPQDOFQK-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-phenyl-1,3,2-dioxaborolane | |
| 570673 | |
| ≥98.0% (GC) |
| 24388-23-6 | |
| 130°C | |
| MFCD00966985 MFCD31699955 | |
| phenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, phenylboronic acid, pinacol ester, 2-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenyl, acmc-209gbq, phenylboronic pinacol ester, phenylboronicacidpinacolester, pinacolboryl benzene | |
| CC1(C)OB(OC1(C)C)C1=CC=CC=C1 | |
| 204.08 | |
| 204.08 | |
| Crystalline Lumps |
Chemical Identifiers
| 24388-23-6 | |
| 204.08 | |
| KKLCYBZPQDOFQK-UHFFFAOYSA-N | |
| 570673 | |
| CC1(C)OB(OC1(C)C)C1=CC=CC=C1 |
| C12H17BO2 | |
| MFCD00966985 MFCD31699955 | |
| phenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, phenylboronic acid, pinacol ester, 2-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenyl, acmc-209gbq, phenylboronic pinacol ester, phenylboronicacidpinacolester, pinacolboryl benzene | |
| 4,4,5,5-tetramethyl-2-phenyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No