missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. p-7135292
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18555G 5 g
P185525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18555G Supplier TCI America Supplier No. P18555G

Chemical Identifiers

CAS 24388-23-6
Molecular Formula C12H17BO2
Molecular Weight (g/mol) 204.08
MDL Number MFCD00966985 MFCD31699955
InChI Key KKLCYBZPQDOFQK-UHFFFAOYSA-N
Synonym phenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, phenylboronic acid, pinacol ester, 2-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenyl, acmc-209gbq, phenylboronic pinacol ester, phenylboronicacidpinacolester, pinacolboryl benzene
PubChem CID 570673
IUPAC Name 4,4,5,5-tetramethyl-2-phenyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1

Specifications

Color White-Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 204.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane