missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-2-(2-pyridyl)acetamide 98.0+%, TCI America™
Supplier: TCI America P174125G
Specifications
| 2-Phenyl-2-(2-pyridyl)acetamide | |
| 136°C | |
| C13H12N2O | |
| 25 g | |
| C1=CC=C(C=C1)C(C2=CC=CC=N2)C(=O)N | |
| 212.252 | |
| 212.25 | |
| Crystalline Powder |
| 7251-52-7 | |
| White-Yellow | |
| MFCD00138126 | |
| PYAPITOPBTXXNJ-UHFFFAOYSA-N | |
| 2-phenyl-2-pyridin-2-ylacetamide | |
| 97964 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 7251-52-7 | |
| 212.252 | |
| PYAPITOPBTXXNJ-UHFFFAOYSA-N | |
| 2-phenyl-2-pyridin-2-ylacetamide |
| C13H12N2O | |
| MFCD00138126 | |
| 97964 | |
| C1=CC=C(C=C1)C(C2=CC=CC=N2)C(=O)N |
Safety and Handling
TSCA : No