missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-(2-pyridyl)acetamide 98.0+%, TCI America™

Catalog No. P17415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17415G 5 g
P174125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17415G Supplier TCI America Supplier No. P17415G
Only null left

Chemical Identifiers

CAS 7251-52-7
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00138126
InChI Key PYAPITOPBTXXNJ-UHFFFAOYSA-N
PubChem CID 97964
IUPAC Name 2-phenyl-2-pyridin-2-ylacetamide
SMILES C1=CC=C(C=C1)C(C2=CC=CC=N2)C(=O)N

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 5 g
Formula Weight 212.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-2-(2-pyridyl)acetamide