missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-propanediol, 98%

Catalog No. AAH5676114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5676114 25 g
AAH5676106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5676114 Supplier Thermo Scientific Chemicals Supplier No. H5676114

CAS: 1570-95-2 | C9H12O2 | 152.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1570-95-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00236056
InChI Key BPBDZXFJDMJLIB-UHFFFAOYSA-N
Synonym 2-phenyl-1,3-propanediol, 2-phenyl-1,3-propanediole, 2-phenyl-propane-1,3-diol, 2-phenyl-1,3-propane diol, unii-9f93674jbb, 1,3-propanediol, 2-phenyl, phenylpropanediol, pubchem20386, rarechem al bd 1370, acmc-1bz5a
PubChem CID 254178
IUPAC Name 2-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(CO)CO

Specifications

Melting Point 53°C to 56°C
Quantity 25 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-Phenyl-1,3-propanediol
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 411-810-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only