missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-1,3-dioxan-5-ol 90.0+%, TCI America™
Supplier: TCI America P142225G
Specifications
| 2-Phenyl-1,3-dioxan-5-ol | |
| 84°C | |
| C10H12O3 | |
| 25 g | |
| BWKDAAFSXYPQOS-UHFFFAOYSA-N | |
| 2-phenyl-1,3-dioxan-5-ol | |
| 74362 | |
| ≥90.0% (NMR) |
| 1708-40-3 | |
| White | |
| MFCD00274343,MFCD00047504 | |
| 1,3-O-Benzylideneglycerol, 5-Hydroxy-2-phenyl-1,3-dioxane | |
| OC1COC(OC1)C1=CC=CC=C1 | |
| 180.20 | |
| 180.20 | |
| Crystalline Powder |
Chemical Identifiers
| 1708-40-3 | |
| 180.20 | |
| BWKDAAFSXYPQOS-UHFFFAOYSA-N | |
| 74362 | |
| OC1COC(OC1)C1=CC=CC=C1 |
| C10H12O3 | |
| MFCD00274343,MFCD00047504 | |
| 1,3-O-Benzylideneglycerol, 5-Hydroxy-2-phenyl-1,3-dioxane | |
| 2-phenyl-1,3-dioxan-5-ol |
Safety and Handling
EINECSNumber : (5)-3571
TSCA : Yes
Recommended Storage : Refrigerator