missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-dioxan-5-ol 90.0+%, TCI America™

Catalog No. p-7135130
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14221G 1 g
P14225G 5 g
P142225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P14221G Supplier TCI America Supplier No. P14221G
Only null left

Chemical Identifiers

CAS 1708-40-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00274343,MFCD00047504
InChI Key BWKDAAFSXYPQOS-UHFFFAOYSA-N
Synonym 1,3-O-Benzylideneglycerol, 5-Hydroxy-2-phenyl-1,3-dioxane
PubChem CID 74362
IUPAC Name 2-phenyl-1,3-dioxan-5-ol
SMILES OC1COC(OC1)C1=CC=CC=C1

Specifications

Melting Point 84°C
Color White
Quantity 1 g
Formula Weight 180.20
Percent Purity ≥90.0% (NMR)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-1,3-dioxan-5-ol
EINECSNumber (5)-3571
TSCA Yes
Recommended Storage Refrigerator