missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxypropylamine, 98%

Catalog No. p-7033580
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50720MD 250 mg
AAH5072003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50720MD Supplier Thermo Scientific Chemicals Supplier No. H50720MD

CAS: 6437-49-6 | C9H14ClNO | 187.667 g/mol

2-Phenoxypropylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxypropylamine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 6437-49-6
Molecular Formula C9H14ClNO
Molecular Weight (g/mol) 187.667
MDL Number MFCD07781046
InChI Key CGOBBGUREQJPPH-UHFFFAOYSA-N
Synonym 2-phenoxypropylamine hydrochloride, 2-phenoxypropan-1-amine hydrochloride, 2-phenoxypropylamine hcl, 1-aminopropan-2-yl oxy benzene hydrochloride, acmc-20annz, 2-phenoxypropan-1-amine-hydrogen chloride 1/1
PubChem CID 24212021
IUPAC Name 2-phenoxypropan-1-amine;hydrochloride
SMILES CC(CN)OC1=CC=CC=C1.Cl

Specifications

Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material 2-Phenoxypropylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only