missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethyl bromide, 98%

Catalog No. AAB2108736
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2108736 500 g
AAB2108714 25 g
AAB2108722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2108736 Supplier Thermo Scientific Chemicals Supplier No. B2108736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 589-10-6 | C8H9BrO | 201.063 g/mol

It is used in the synthesis and antihypertensive activity of 4'-substituted spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-ones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and antihypertensive activity of 4′-substituted spiro[4H-3,1-benzoxazine-4,4′-piperidin]-2(1H)-ones.

Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 589-10-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00000234
InChI Key JJFOBACUIRKUPN-UHFFFAOYSA-N
Synonym 2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene
PubChem CID 68526
IUPAC Name 2-bromoethoxybenzene
SMILES C1=CC=C(C=C1)OCCBr

Specifications

Melting Point 31°C to 35°C
Density 1.35
Boiling Point 135°C to 137°C (20 mmHg)
Flash Point 65°C (149°F)
Quantity 500 g
Beilstein 508290
Solubility Information Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.
Formula Weight 201.07
Percent Purity 98%
Chemical Name or Material 2-Phenoxyethyl bromide
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 209-634-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only