missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxyethanol 98.5+%, TCI America™
Supplier: TCI America P0115500G
Specifications
| 2-Phenoxyethanol | |
| Colorless | |
| C8H10O2 | |
| 500 g | |
| QCDWFXQBSFUVSP-UHFFFAOYSA-N | |
| 2-phenoxyethanol | |
| 31236 | |
| 138.17 | |
| Liquid |
| 122-99-6 | |
| 237°C | |
| MFCD00002857 | |
| phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane | |
| C1=CC=C(C=C1)OCCO | |
| 138.166 | |
| CHEBI:64275 | |
| ≥98.5% (GC) |
Chemical Identifiers
| 122-99-6 | |
| 138.166 | |
| QCDWFXQBSFUVSP-UHFFFAOYSA-N | |
| 31236 | |
| 2-phenoxyethanol |
| C8H10O2 | |
| MFCD00002857 | |
| phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane | |
| CHEBI:64275 | |
| C1=CC=C(C=C1)OCCO |
Safety and Handling
EINECSNumber : (3)-0558&(9)-1277&(7)-0078
RTECSNumber : KM0350000
TSCA : Yes