missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol 98.5+%, TCI America™

Catalog No. P011525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P011525G 25 g
P0115500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P011525G Supplier TCI America Supplier No. P011525G

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002857
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
Synonym phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
PubChem CID 31236
ChEBI CHEBI:64275
IUPAC Name 2-phenoxyethanol
SMILES C1=CC=C(C=C1)OCCO

Specifications

Color Colorless
Boiling Point 237°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethanol
EINECSNumber (3)-0558&(9)-1277&(7)-0078
RTECSNumber KM0350000
TSCA Yes