missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol 98.5+%, TCI America™

Supplier:  TCI America P195325G

 View more versions of this product

Catalog No. P195325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Phenoxyethanol [for Biochemical Research]
Colorless
C8H10O2
25 g
QCDWFXQBSFUVSP-UHFFFAOYSA-N
2-phenoxyethanol
31236
138.17
Liquid
122-99-6
237°C
MFCD00002857
phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
C1=CC=C(C=C1)OCCO
138.166
CHEBI:64275
≥98.5% (GC)

Chemical Identifiers

122-99-6
138.166
QCDWFXQBSFUVSP-UHFFFAOYSA-N
31236
2-phenoxyethanol
C8H10O2
MFCD00002857
phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
CHEBI:64275
C1=CC=C(C=C1)OCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (7)-0078&(3)-0558&(9)-1277

RTECSNumber : KM0350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions