Learn More
2-Phenoxybenzoyl chloride, TECH, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC49902DA
Specifications
| 2-Phenoxybenzoyl chloride | |
| 89.5 | |
| 90% | |
| MFCD03424711 | |
| benzoylchloride, 2-phenoxy, phenoxybenzoyl chloride, 2-phenoxy benzoyl chloride, 2-phenoxy-benzoyl chloride, benzoyl chloride, 2-phenoxy, 2-phenoxybenzoic acid chloride, 2-chlorocarbonyl diphenyl ether | |
| C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)Cl | |
| 232.663 | |
| 232.67 |
| 40501-36-8 | |
| 100.0 | |
| C13H9ClO2 | |
| 1g | |
| BMGKQFRMINVVPP-UHFFFAOYSA-N | |
| 2-phenoxybenzoyl chloride | |
| 11075303 | |
| 90% |
Chemical Identifiers
| 40501-36-8 | |
| 232.663 | |
| BMGKQFRMINVVPP-UHFFFAOYSA-N | |
| 11075303 | |
| C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)Cl |
| C13H9ClO2 | |
| MFCD03424711 | |
| benzoylchloride, 2-phenoxy, phenoxybenzoyl chloride, 2-phenoxy benzoyl chloride, 2-phenoxy-benzoyl chloride, benzoyl chloride, 2-phenoxy, 2-phenoxybenzoic acid chloride, 2-chlorocarbonyl diphenyl ether | |
| 2-phenoxybenzoyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger