missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzoyl chloride, TECH, Thermo Scientific™

Catalog No. p-3175899
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC49902CB 250mg
CC49902DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC49902CB Supplier Thermo Scientific Chemicals Supplier No. CC49902CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40501-36-8
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.663
MDL Number MFCD03424711
InChI Key BMGKQFRMINVVPP-UHFFFAOYSA-N
Synonym benzoylchloride, 2-phenoxy, phenoxybenzoyl chloride, 2-phenoxy benzoyl chloride, 2-phenoxy-benzoyl chloride, benzoyl chloride, 2-phenoxy, 2-phenoxybenzoic acid chloride, 2-chlorocarbonyl diphenyl ether
PubChem CID 11075303
IUPAC Name 2-phenoxybenzoyl chloride
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 232.67
Percent Purity 90%
Chemical Name or Material 2-Phenoxybenzoyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.