missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzoic Acid, TCI America™

Catalog No. P125225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P125225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P125225G Supplier TCI America Supplier No. P125225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2243-42-7
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002429
InChI Key PKRSYEPBQPFNRB-UHFFFAOYSA-N
Synonym o-phenoxybenzoic acid, benzoic acid, 2-phenoxy, benzoic acid, o-phenoxy, phenoxybenzoic acid, ortho-phenoxybenzoic acid, 2-phenoxybenzoicacid, 2-carboxydiphenyl ether, 2-phenoxy-benzoic acid, o-phenoxybenzoic acid, o-carboxydiphenyl ether
PubChem CID 75237
ChEBI CHEBI:72636
IUPAC Name 2-phenoxybenzoic acid
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)O

Specifications

Melting Point 113°C
Color White
Boiling Point 355°C
Quantity 25 g
Formula Weight 214.22
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenoxybenzoic Acid
RTECSNumber DH6293560
TSCA Yes