missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxybenzaldehyde 96.0+%, TCI America™
$113.59 - $342.69
Chemical Identifiers
| CAS | 19434-34-5 |
|---|---|
| Molecular Formula | C13H10O2 |
| Molecular Weight (g/mol) | 198.22 |
| MDL Number | MFCD00800666 |
| InChI Key | IMPIIVKYTNMBCD-UHFFFAOYSA-N |
| Synonym | o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384 |
| PubChem CID | 88060 |
| IUPAC Name | 2-phenoxybenzaldehyde |
| SMILES | O=CC1=CC=CC=C1OC1=CC=CC=C1 |
Chemical Identifiers
| 19434-34-5 | |
| 198.22 | |
| IMPIIVKYTNMBCD-UHFFFAOYSA-N | |
| 88060 | |
| O=CC1=CC=CC=C1OC1=CC=CC=C1 |
| C13H10O2 | |
| MFCD00800666 | |
| o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384 | |
| 2-phenoxybenzaldehyde |
Specifications
| 19434-34-5 | |
| 153°C | |
| MFCD00800666 | |
| o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384 | |
| O=CC1=CC=CC=C1OC1=CC=CC=C1 | |
| 198.22 | |
| 198.22 | |
| Liquid |
| Yellow | |
| C13H10O2 | |
| 1 g | |
| IMPIIVKYTNMBCD-UHFFFAOYSA-N | |
| 2-phenoxybenzaldehyde | |
| 88060 | |
| ≥96.0% (GC) | |
| 2-Phenoxybenzaldehyde |
Safety and Handling
TSCA : No