missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzaldehyde 96.0+%, TCI America™

Catalog No. P24755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24755G 5 g
P24751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24755G Supplier TCI America Supplier No. P24755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19434-34-5
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00800666
InChI Key IMPIIVKYTNMBCD-UHFFFAOYSA-N
Synonym o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384
PubChem CID 88060
IUPAC Name 2-phenoxybenzaldehyde
SMILES O=CC1=CC=CC=C1OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxybenzaldehyde