missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyacetophenone 98.0+%, TCI America™

Catalog No. P21215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21215G 5 g
P21211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21215G Supplier TCI America Supplier No. P21215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 721-04-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00156689
InChI Key KRSXGTAVHIDVPM-UHFFFAOYSA-N
PubChem CID 222171
ChEBI CHEBI:52401
IUPAC Name 2-phenoxy-1-phenylethan-1-one
SMILES O=C(COC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 73°C
Color White-Yellow
Boiling Point 178°C
Quantity 5 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenoxyacetophenone