accessibility menu, dialog, popup

UserName

CAS RN 721-04-0

IUPAC Name: 2-phenoxy-1-phenylethan-1-one
Synonyms: 2-phenoxyacetophenone
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: KRSXGTAVHIDVPM-UHFFFAOYSA-N
SMILES: O=C(COC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results